missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako PYROSTAR™ ES-F 80 Test Kit

Catalog No. 12333000
Change view
Click to view available options
Sensitivity:
0.015 EU/mL
0.03 EU/mL
0.06 EU/mL
0.125 EU/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensitivity
12333000 0.015 EU/mL
12333001 0.03 EU/mL
12333002 0.06 EU/mL
12333003 0.125 EU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12333000 Supplier FUJIFILM Wako Supplier No. WPEK420015

Endotoxin-specific and specially formulated to be unreactive to (1,3)- β-D-glucan, Multi-test configuration designed for use with water bath, heat block or Toxinometer™

  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Gel clot sensitivities range from 0.015 to 0.25 EU/mL
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Chances of interference are reduced due to high sensitivity
  • Facilitates easier transition from gel clot to KTA

Specifications

Form Lyophilized
Fill Volume 2 mL
Includes 4 x 2 mL multi-test vials and 1 vial CSE (500 ng/vial)
Quantity 80 Tests
Sensitivity 0.015 EU/mL