missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic Dianhydride (purified by sublimation) 99.0+%, TCI America™

Catalog No. P21035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21035G 5 g
P210325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21035G Supplier TCI America Supplier No. P21035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-32-7
Molecular Formula C10H2O6
Molecular Weight (g/mol) 218.12
MDL Number MFCD00005005
InChI Key ANSXAPJVJOKRDJ-UHFFFAOYSA-N
Synonym pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydride
PubChem CID 6966
IUPAC Name 5,11-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),8-triene-4,6,10,12-tetrone
SMILES O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O

Specifications

Melting Point 286°C
Color White
Boiling Point 400°C
Quantity 5 g
Formula Weight 218.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyromellitic Dianhydride (purified by sublimation)
EINECSNumber (3)-1287
RTECSNumber DB9300000
TSCA Yes