missing translation for 'onlineSavingsMsg'
Learn More

Pyromellitic Acid 98.0+%, TCI America™

Catalog No. B003925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003925G 25 g
B0039100G 100 g
B0039500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B003925G Supplier TCI America Supplier No. B003925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-05-4 | C10H6O8 | 254.15 g/mol

Chemical Identifiers

CAS 89-05-4
Molecular Formula C10H6O8
Molecular Weight (g/mol) 254.15
MDL Number MFCD00002471
InChI Key CYIDZMCFTVVTJO-UHFFFAOYSA-N
Synonym pyromellitic acid, 1,2,4,5-benzenetetracarboxylic acid, usaf xr-20, 1,2,4,5-tetracarboxybenzene, unii-d9uxg2u6zu, 1,2,4,5-benzene-tetracarboxylic acid, d9uxg2u6zu, pubchem21809, acmc-20ap78, 1,4,5-tetracarboxybenzene
PubChem CID 6961
ChEBI CHEBI:45165
IUPAC Name benzene-1,2,4,5-tetracarboxylic acid
SMILES C1=C(C(=CC(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 286°C
Color White
Quantity 25 g
Formula Weight 254.15
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Pyromellitic Acid
EINECSNumber (3)-2950
RTECSNumber DB9275000
TSCA Yes