missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol Red, TCI America™

Catalog No. P05721G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P05721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P05721G Supplier TCI America Supplier No. P05721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32638-88-3
Molecular Formula C19H11O8S
Molecular Weight (g/mol) 399.35
MDL Number MFCD00005046
InChI Key JTZKNOUMMWIARI-UHFFFAOYSA-M
Synonym pyrogallol red, pyrogallol sulfonphthalein, 3',4',5',6'-tetrahydroxyspiro benzo c 1,2 oxathiole-3,9'-xanthene 1,1-dioxide, spiro 3h-2,1-benzoxathiole-3,9'-9h xanthene-3',4',5',6'-tetrol, 1,1-dioxide, us8987474, pyrogallolred, pyrogallolred forproteinresearch, 12,13,14,15-tetrahydroxyspiro benzo c 1,2-oxathiolane-3,9'-xanthene-1,1-dione, 3',4',5',6'-tetrahydroxyspiro 2,1$l^ 6-benzoxathiole-3,9'-xanthene-1,1-dione, 3',4',5',6'-tetrahydroxyspiro 2,1??-benzoxathiole-3,9'-xanthene-1,1-dione
PubChem CID 94437
IUPAC Name 2-(4,5,6-trihydroxy-3-oxo-3H-xanthen-9-yl)benzene-1-sulfonate
SMILES OC1=CC=C2C(OC3=C(O)C(=O)C=CC3=C2C2=CC=CC=C2S([O-])(=O)=O)=C1O

Specifications

Quantity 1 g
Formula Weight 400.36
Physical Form Crystalline Powder
Chemical Name or Material Pyrogallol Red