missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrogallol Red (Ready-to-use solution) [for Protein determination], TCI America™
Specifications
Specifications
| Color | Red-Yellow |
| Quantity | 100 mL |
| UN Number | 1993 |
| Physical Form | Liquid |
| Chemical Name or Material | Pyrogallol Red (Ready-to-use solution) [for Protein determination] |
Safety and Handling
| Recommended Storage | Refrigerator |