missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-188735
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A263100 100 g Glass Bottle
A263500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A263100 Supplier Thermo Fisher Scientific Supplier No. FLA263100
Only null left

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131°C
Color White
Density 1.453g/cm³
pH 5.8
Boiling Point 309°C
Ignition Residue 0.005% max.
Melting Range 131°C to 135°C
Packaging Glass Bottle
Linear Formula C6H3-1, 2, 3-(OH)3
Quantity 100 g
Formula Weight 126.11
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Pyrogallol
Show More Show Less
DOTInformation DOT Class 6.1, : Poison