missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A263100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A263100 100 g Glass Bottle
A263500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A263100 Supplier Thermo Fisher Scientific Supplier No. FLA263100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131°C
Color White
Density 1.453g/cm³
pH 5.8
Boiling Point 309°C
Ignition Residue 0.005% max.
Melting Range 131°C to 135°C
Packaging Glass Bottle
Linear Formula C6H3-1, 2, 3-(OH)3
Quantity 100 g
Formula Weight 126.11
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Pyrogallol
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison