missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, ACS reagent

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

$329.61 - $329.61

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC418621000
SDS View Documents
Thermo Scientific Chemicals
418621000
100 g Plastic bottle
Each for $329.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

87-66-1
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
1057
benzene-1,2,3-triol
C6H6O3
MFCD00002192
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
CHEBI:16164
OC1=CC=CC(O)=C1O

Specifications

87-66-1
White
0.005% max.
Plastic bottle
C6H3(OH)3
100 g
06, 1071
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
1057
126.11
Crystalline Powder
131.0°C to 135.0°C
309.0°C
Authentic
C6H6O3
MFCD00002192
2811
15, 8112
Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide
OC1=CC=CC(O)=C1O
126.11
CHEBI:16164
ACS Reagent
Pyrogallol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Suspected of causing genetic defects.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serio

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr

GHS Signal Word: Danger

EINECSNumber : 201-762-9

RTECSNumber : UX2800000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only