missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, 99%

Catalog No. AC164561000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164561000 100 g Plastic bottle
AC164565000 500 g Plastic bottle
AC164560025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164561000 Supplier Thermo Scientific Chemicals Supplier No. 164561000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131.0°C to 135.0°C
Color White
Boiling Point 309.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H3(OH)3
Quantity 100 g
Beilstein 06,1071
Merck Index 15,8112
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 1 g/1.3 mL alcohol, 1 g/1.6 mL ether, slightly soluble in benzene, chloroform and, carbon disulfide
Formula Weight 126.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Pyrogallol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Toxic if swallowed.
Suspected of causing genetic defects.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serio
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irr
EINECSNumber 201-762-9

RUO – Research Use Only