missing translation for 'onlineSavingsMsg'
Learn More

Pyrogallol, 98+%

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

Supplier:  Thermo Scientific Chemicals A1340522

 View more versions of this product

Catalog No. AAA1340522


Only null left
Explore available promotions

Description

Description

Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

Solubility
Soluble in ethanol, water and ether. Slightly soluble in chloroform.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkalis, metal oxides, ammonia, iodine, lime water, potassium permanganate and strong bases.
Specifications

Specifications

Pyrogallol
131°C to 135°C
309°C
C6H6O3
100 g
907431
14,8000
Soluble in ethanol,water and ether. Slightly soluble in chloroform.
OC1=CC=CC(O)=C1O
126.11
CHEBI:16164
≥98%
87-66-1
1.45
Odorless
MFCD00002192
UN2811
Air and light sensitive
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
1057
126.11

Chemical Identifiers

87-66-1
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
1057
benzene-1,2,3-triol
C6H6O3
MFCD00002192
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
CHEBI:16164
OC1=CC=CC(O)=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H312-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H301-H312+H332-H315-H319-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 201-762-9

RTECSNumber : UX2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only