missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxylic Acid 97.0+%, TCI America™

Catalog No. P19091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19091G Supplier TCI America Supplier No. P19091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4595-61-3
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856162
InChI Key IIVUJUOJERNGQX-UHFFFAOYSA-N
Synonym 5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
PubChem CID 78346
IUPAC Name pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=CN=C1

Specifications

Melting Point 270°C
Quantity 1 g
Formula Weight 124.10
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrimidine-5-carboxylic Acid