missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-carboxaldehyde, 97%

Catalog No. p-7036193
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5444803 1 g
AAH5444806 5 g
AAH5444814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5444803 Supplier Thermo Scientific Chemicals Supplier No. H5444803

CAS: 10070-92-5 | C5H4N2O | 108.10 g/mol

Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10070-92-5
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.10
MDL Number MFCD03426065
InChI Key FREJAOSUHFGDBW-UHFFFAOYSA-N
Synonym pyrimidine-5-carboxaldehyde, 5-formylpyrimidine, pyrimidine-5-carboxyaldehyde, 5-pyrimidinecarboxaldehyde, 5-pyrimidinecarbaldehyde, 5-pyrimidinaldehyde, 5-formyl pyrimidine, pubchem5298, pyrimidine-5-aldehyde, pyrimidine 5-carboxaldehyde
PubChem CID 2761034
IUPAC Name pyrimidine-5-carbaldehyde
SMILES O=CC1=CN=CN=C1

Specifications

Melting Point 39°C to 43°C
Density 1.23
Boiling Point 80°C to 82°C (4 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 108.1
Percent Purity 97%
Chemical Name or Material Pyrimidine-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only