missing translation for 'onlineSavingsMsg'
Learn More

Pyrimidine-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH2714703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2714703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2714703 Supplier Thermo Scientific Chemicals Supplier No. H2714703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 321724-19-0 | Molecular Formula: C10H15BN2O2 | Molecular Weight: 206.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 321724-19-0
Molecular Formula C10H15BN2O2
Molecular Weight (g/mol) 206.05
MDL Number MFCD05155223
InChI Key WSRGGSAGSUEJKQ-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine-5-boronic acid pinacol ester, pinacol ester pyrimidinyl-5-boronic acid, 5-pyrimidineboronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidyl-5-boronic acid pinacolate, pyrimidine-5-boronic acid, pinacol ester, pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pyrimidin-5-ylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-pyrimidin-5-yl-1,3,2-dioxaborolane
PubChem CID 4192667
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
SMILES CC1(C)OB(OC1(C)C)C1=CN=CN=C1

Specifications

Melting Point 60°C to 66°C
Quantity 1 g
Formula Weight 206.05
Percent Purity 95%
Chemical Name or Material Pyrimidine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only