missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P0561500G
Specifications
| Pyridoxine Hydrochloride | |
| 207°C | |
| C8H12ClNO3 | |
| 500 g | |
| ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
| 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride | |
| 6019 | |
| 205.64 | |
| Crystalline Powder |
| 58-56-0 | |
| White-Yellow | |
| MFCD00012807 | |
| pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
| CC1=NC=C(C(=C1O)CO)CO.Cl | |
| 205.638 | |
| CHEBI:30961 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 58-56-0 | |
| 205.638 | |
| ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
| 6019 | |
| 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride |
| C8H12ClNO3 | |
| MFCD00012807 | |
| pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
| CHEBI:30961 | |
| CC1=NC=C(C(=C1O)CO)CO.Cl |
Safety and Handling
EINECSNumber : (9)-1043
RTECSNumber : UV1350000
TSCA : Yes