missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Hydrochloride, FCC, 98-100.5%, Spectrum™ Chemical

Catalog No. p-7158121
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605989 25 g Amber Glass Bottle
18605990 100 g Amber Glass Bottle
18605991 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605989 Supplier Spectrum Chemical Mfg Cor Supplier No. P145925GM
Only null left

P1459, 58-56-0, C8H11NO3·HCl

Spectrum™ Chemical Pyridoxine Hydrochloride, FCC , a form of vitamin B6, is used as a vitamin B6 dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.64
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
SMILES [H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 100.5%
Grade FCC