missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine hydrochloride, 99%

CAS: 58-56-0 | C8H12ClNO3 | 205.638 g/mol

Supplier:  Thermo Scientific Chemicals A1204122

 View more versions of this product

Catalog No. AAA1204122


Only null left
Explore available promotions

Description

Description

Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.

Solubility
Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.

Notes
Light sensitive. Incompatible with bases and strong oxidizing agents.
Specifications

Specifications

Pyridoxine hydrochloride
∼206°C (decomposition)
Odorless
MFCD00012807
3632435
14,7982
Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.
CC1=NC=C(C(=C1O)CO)CO.Cl
205.638
CHEBI:30961
99%
58-56-0
3
C8H12ClNO3
100 g
Light sensitive
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
6019
205.64

Chemical Identifiers

58-56-0
205.638
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
C8H12ClNO3
MFCD00012807
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CHEBI:30961
CC1=NC=C(C(=C1O)CO)CO.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-386-2

RTECSNumber : UV1350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only