missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pyridoxine hydrochloride, 98+%, extra pure

CAS: 58-56-0 | C8H11NO3·HCl | 205.64 g/mol

Supplier:  Thermo Scientific Chemicals 150772500

 View more versions of this product

Catalog No. AC150772500

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pyridoxine hydrochloride
214.0°C
2.4 to 3.0 (5% aq. soln.)
Authentic
Glass bottle
MFCD00012807
21, V,5, 492
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
6019
205.64
Extra Pure
58-56-0
White
0.5% max. (105°C)
98+%
C8H11NO3·HCl
250 g
15, 8095
Solution
CC1=NC=C(C(=C1O)CO)CO.Cl
205.64
CHEBI:30961
98+%
Crystalline Powder

Chemical Identifiers

58-56-0
205.64
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
C8H11NO3·HCl
MFCD00012807
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CHEBI:30961
CC1=NC=C(C(=C1O)CO)CO.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-386-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only