missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridoxine Dicaprylate 98.0+%, TCI America™
$78.29 - $231.14
Chemical Identifiers
| CAS | 106483-04-9 |
|---|---|
| Molecular Formula | C24H39NO5 |
| Molecular Weight (g/mol) | 421.58 |
| MDL Number | MFCD28386112 |
| InChI Key | PAUSGZCRNOTKPK-UHFFFAOYSA-N |
| PubChem CID | 83816 |
| IUPAC Name | {3-hydroxy-2-methyl-5-[(octanoyloxy)methyl]pyridin-4-yl}methyl octanoate |
| SMILES | CCCCCCCC(=O)OCC1=CN=C(C)C(O)=C1COC(=O)CCCCCCC |
Chemical Identifiers
| 106483-04-9 | |
| 421.58 | |
| PAUSGZCRNOTKPK-UHFFFAOYSA-N | |
| {3-hydroxy-2-methyl-5-[(octanoyloxy)methyl]pyridin-4-yl}methyl octanoate |
| C24H39NO5 | |
| MFCD28386112 | |
| 83816 | |
| CCCCCCCC(=O)OCC1=CN=C(C)C(O)=C1COC(=O)CCCCCCC |
Specifications
| 106483-04-9 | |
| White | |
| MFCD28386112 | |
| PAUSGZCRNOTKPK-UHFFFAOYSA-N | |
| {3-hydroxy-2-methyl-5-[(octanoyloxy)methyl]pyridin-4-yl}methyl octanoate | |
| 83816 | |
| ≥98.0% (N) | |
| Pyridoxine Dicaprylate |
| 72°C | |
| C24H39NO5 | |
| 5 g | |
| CCCCCCCC(=O)OCC1=CN=C(C)C(O)=C1COC(=O)CCCCCCC | |
| 421.58 | |
| 421.58 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-2209
TSCA : No