missing translation for 'onlineSavingsMsg'
Learn More

Pyridostigmine Bromide 98.0+%, TCI America™

Catalog No. p-7135085
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13395G 5 g
P133925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13395G Supplier TCI America Supplier No. P13395G
Only null left

Chemical Identifiers

CAS 101-26-8
Molecular Formula C9H13BrN2O2
Molecular Weight (g/mol) 261.12
MDL Number MFCD00079283
InChI Key VNYBTNPBYXSMOO-UHFFFAOYSA-M
Synonym pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide
PubChem CID 7550
IUPAC Name 3-[(dimethylcarbamoyl)oxy]-1-methylpyridin-1-ium bromide
SMILES [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1

Specifications

Melting Point 154°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 261.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridostigmine Bromide
EINECSNumber (9)-0648
RTECSNumber UU5270000
TSCA No