Learn More
Pyridostigmine bromide, 98%
CAS: 101-26-8 | C9H13BrN2O2 | 261.12 g/mol
$136.41 - $136.41
Chemical Identifiers
| CAS | 101-26-8 |
|---|---|
| Molecular Formula | C9H13BrN2O2 |
| Molecular Weight (g/mol) | 261.12 |
| MDL Number | MFCD00079283 |
| InChI Key | VNYBTNPBYXSMOO-UHFFFAOYSA-M |
| Synonym | pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide |
| PubChem CID | 7550 |
| IUPAC Name | (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide |
| SMILES | [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461190010
|
Thermo Scientific Chemicals
461190010 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 101-26-8 | |
| 261.12 | |
| VNYBTNPBYXSMOO-UHFFFAOYSA-M | |
| 7550 | |
| [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1 |
| C9H13BrN2O2 | |
| MFCD00079283 | |
| pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide | |
| (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide |
Specifications
| 101-26-8 | |
| 2% max. (1 g, 105°C) | |
| 98% | |
| C9H13BrN2O2 | |
| 1 g | |
| pyridostigmine bromide, mestinon, regonol, kalimin, kalymin, regonal, 3-dimethylcarbamoyl oxy-1-methylpyridin-1-ium bromide, mestinon bromide, 3-dimethylcarbamoyloxy-1-methylpyridinium bromide | |
| VNYBTNPBYXSMOO-UHFFFAOYSA-M | |
| (1-methylpyridin-1-ium-3-yl) N,N-dimethylcarbamate;bromide | |
| 7550 | |
| 98% |
| 154.0°C | |
| Authentic | |
| Glass Bottle | |
| MFCD00079283 | |
| 15,809 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, chloroform, insoluble in ether, benzene, acetone | |
| [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1 | |
| 261.12 | |
| 261.12 | |
| Pyridostigmine bromide |
Safety and Handling
GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. May cause an allergic skin reaction.
GHS P Statement Rinse mouth. Wear protective gloves. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Danger
EINECSNumber : 202-929-9
RUO – Research Use Only