missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium Dichromate 98.0+%, TCI America™

Catalog No. P093125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P093125G 25 g
P0931500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P093125G Supplier TCI America Supplier No. P093125G

Chemical Identifiers

CAS 20039-37-6
Molecular Formula C10H12Cr2N2O7
Molecular Weight (g/mol) 376.205
MDL Number MFCD00013105
InChI Key LMYWWPCAXXPJFF-UHFFFAOYSA-P
Synonym pyridinium dichromate, cornforth reagent, pyridinium dichromate pdc, oxido-oxido dioxo chromio oxy-dioxochromium; pyridin-1-ium, pyridiniumdichromate, pyridinium-dichromate, dipyridinium dichromate, bis pyridium chromate, pyridiniumdichromate pdc, acmc-209f5g
PubChem CID 2724130
IUPAC Name oxido-(oxido(dioxo)chromio)oxy-dioxochromium;pyridin-1-ium
SMILES C1=CC=[NH+]C=C1.C1=CC=[NH+]C=C1.[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 149°C
Quantity 25 g
UN Number 1479
Formula Weight 376.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Dichromate