missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium Chlorochromate 98.0+%, TCI America™

Catalog No. P093025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P093025G 25 g
P0930100G 100 g
P0930500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P093025G Supplier TCI America Supplier No. P093025G

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H6ClCrNO3
MDL Number MFCD00013106
Synonym PCC

Specifications

Quantity 25 g
UN Number 3087
Formula Weight 215.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium Chlorochromate