missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium chlorochromate, 98%

Catalog No. p-189712
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183670250 25 g Plastic bottle
AC183671000 100 g Plastic bottle
AC183675000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC183670250 Supplier Thermo Scientific Chemicals Supplier No. 183670250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26299-14-9 | C5H6ClCrNO3 | 215.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26299-14-9
Molecular Formula C5H6ClCrNO3
Molecular Weight (g/mol) 215.56
MDL Number MFCD00013106
Synonym PCC

Specifications

Melting Point 205.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Fieser 06,498; 07,308; 08,425; 09,397; 10,334; 11,450; 12,256; 13,237; 14,269; 15,57
Merck Index 15, 8086
Solubility Information Solubility in water: .. Other solubilities: insoluble in dichloromethane
Formula Weight 215.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium chlorochromate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 247-595-5
TSCA TSCA

RUO – Research Use Only