missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium 3-Nitrobenzenesulfonate 98.0+%, TCI America™

Catalog No. p-7134860
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09415G 5 g
P094125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09415G Supplier TCI America Supplier No. P09415G
Only null left

Chemical Identifiers

CAS 84752-61-4
Molecular Formula C11H10N2O5S
Molecular Weight (g/mol) 282.27
MDL Number MFCD00060234
InChI Key WZZFZXZRKPNZOC-UHFFFAOYSA-N
PubChem CID 16211770
IUPAC Name 3-nitrobenzenesulfonate;pyridin-1-ium
SMILES C1=CC=[NH+]C=C1.C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridinium 3-Nitrobenzenesulfonate