missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 55 gal. |
| Packaging | Drum |
| Boiling Point | 115°C |
| Grade | Technical |
| Chemical Name or Material | Pyridine |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.