missing translation for 'onlineSavingsMsg'
Learn More

Pyridine 99.0+%, TCI America™

Catalog No. Q0034100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0034100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q0034100ML Supplier TCI America Supplier No. Q0034100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42°C
Color Colorless
pH 8.5 (0.2 mol H2O soln.)
Boiling Point 115°C
Quantity 100 mL
UN Number 1282
Formula Weight 79.10
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyridine [Sequencing Solvent]
EINECSNumber (5)-0710
RTECSNumber UR8400000
TSCA Yes