missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7158119
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605973 100 mL Amber Glass Bottle
18605975 500 mL Amber Glass Bottle
18605977 1 L Amber Glass Bottle
18605976 6 x 500 mL Amber Glass Bottle
18605980 4 L Amber Glass Bottle
18605978 6 x 1 L Amber Glass Bottle
18605981 4 x 4 L Amber Glass Bottle
18605974 20 L Steel Can
18605979 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18605973 Supplier Spectrum Chemical Mfg Cor Supplier No. P1450100ML
Only null left

P1450, 110-86-1, C5H5N

Spectrum™ Chemical Pyridine, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

CAS Max % 1
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 2e-005
Water 0.001
Boiling Point 115°C
Percent Purity 99%
Grade Reagent
Chemical Name or Material Pyridine