missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, Reagent, ACS, 99%, Spectrum™ Chemical
Description
Spectrum™ Chemical Pyridine, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 200 L |
| Packaging | Drum |
| Residue after Evaporation | 2e-005 |
| Water | 0.001 |
| Boiling Point | 115°C |
| Percent Purity | 99% |
| Grade | Reagent |
| Chemical Name or Material | Pyridine |