missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, puriss., Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120743
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046507 1 L Glass Bottle
60046508 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046507 Supplier Honeywell Chemicals Supplier No. 349451L
Only null left

Puriss., Reag. Ph. Eur., dried, ≤0.0075% water,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH ∼8.8 (at 20°C)
Melting Point -42°C
Color Colorless
Density 0.978g/mL at 25°C
Boiling Point 115°C
Quantity 1 L
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Odor Unpleasant
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 10mmHg (13.2°C), 20mmHg (25°C)
UN Number UN1282
Beilstein 103233
Solubility Information Completely miscible in water
Formula Weight 79.1g/mol
Percent Purity ≥99.5% (GC)
Grade Reagent Ph. Eur./puriss.
Chemical Name or Material Pyridine
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Pyridine
RTECSNumber 203-809-9