missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120742
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046283 500 mL Glass Bottle
60046282 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046283 Supplier Honeywell Chemicals Supplier No. 33553500ML
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42°C
Color Colorless
Density 0.978g/mL at 25°C
Boiling Point 115°C
Quantity 500 mL
Explosion Limit 0.124
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.509
Vapor Pressure 20mmHg (25°C), 10mmHg (13.2°C)
UN Number UN1282
Beilstein 103233
Formula Weight 79.1g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/Reagent Ph. Eur./puriss. p.a.
Chemical Name or Material Pyridine
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Pyridine
RTECSNumber 203-809-9