missing translation for 'onlineSavingsMsg'
Learn More

Pyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7134675
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
P055725G 25 g
P0557100G 100 g
P0557500G 500 g
3 options

Catalog No. P055725G

Supplier: TCI America P055725G

Only null left

Chemical Identifiers

CAS 694-59-7
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00006194
InChI Key ILVXOBCQQYKLDS-UHFFFAOYSA-N
Synonym pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide
PubChem CID 12753
ChEBI CHEBI:29136
IUPAC Name 1-oxidopyridin-1-ium
SMILES C1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 62°C
Color White
Boiling Point 105°C
Quantity 25 g
Formula Weight 95.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine N-Oxide
EINECSNumber (9)-1106
RTECSNumber UT6410000
TSCA Yes