Learn More
Pyridine N-oxide, 95%
CAS: 694-59-7 | C5H5NO | 95.101 g/mol
Supplier: Thermo Scientific Chemicals A1041922
Description
Pyridine N-oxide is used in the treatment of Kayser's disease. It is widely used in chemical industry. It can act as catalytic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water.
Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Specifications
| Pyridine N-oxide | |
| 61°C to 66°C | |
| 143°C (289°F) | |
| MFCD00006194 | |
| 105257 | |
| 14,7972 | |
| Soluble in water. | |
| C1=CC=[N+](C=C1)[O-] | |
| 95.101 | |
| CHEBI:29136 | |
| 95% |
| 694-59-7 | |
| 270°C | |
| C5H5NO | |
| 100 g | |
| Very Hygroscopic | |
| pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide | |
| ILVXOBCQQYKLDS-UHFFFAOYSA-N | |
| 1-oxidopyridin-1-ium | |
| 12753 | |
| 95.1 |
Chemical Identifiers
| 694-59-7 | |
| 95.101 | |
| ILVXOBCQQYKLDS-UHFFFAOYSA-N | |
| 12753 | |
| 1-oxidopyridin-1-ium |
| C5H5NO | |
| MFCD00006194 | |
| pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide | |
| CHEBI:29136 | |
| C1=CC=[N+](C=C1)[O-] |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 211-774-6
RTECSNumber : UT6410000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only