missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrochloride, 98%, pure

Catalog No. AC222281000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222281000 100 g Plastic bottle
AC222285000 500 g Plastic bottle
AC222280025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222281000 Supplier Thermo Scientific Chemicals Supplier No. 222281000

CAS: 628-13-7 | C5H5N·ClH | 115.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-13-7
Molecular Formula C5H5N·ClH
Molecular Weight (g/mol) 115.56
InChI Key AOJFQRQNPXYVLM-UHFFFAOYSA-N
Synonym pyridine hydrochloride, pyridine, hydrochloride, pyridinehydrochloride, pyridine hcl, pyridine, hydrochloride 1:1, pyridine-hcl, pyridine.hcl, pyridiniumchloride, pyridine hcl salt, pyrdine hydrochloride
PubChem CID 10176127
IUPAC Name pyridine;hydrochloride
SMILES C1=CC=NC=C1.Cl

Specifications

Melting Point 140.0°C to 146.0°C
Color Tan to White
Boiling Point 222.0°C to 224.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Quantity 100 g
Beilstein 20,189
Fieser 01,964; 02,352; 03,239; 04,415
Solubility Information (5% in water) Clear colorless to almost colorless
Formula Weight 115.56
Percent Purity 98%
Grade Pure
Physical Form Crystals
Chemical Name or Material Pyridine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 211-027-4

RUO – Research Use Only