missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine Hydrobromide 98.0+%, TCI America™
$32.72 - $32.72
Chemical Identifiers
| CAS | 18820-82-1 |
|---|---|
| Molecular Formula | C5H6BrN |
| Molecular Weight (g/mol) | 160.014 |
| MDL Number | MFCD00012801 |
| InChI Key | BBFCIBZLAVOLCF-UHFFFAOYSA-N |
| Synonym | pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at |
| PubChem CID | 87810 |
| IUPAC Name | pyridine;hydrobromide |
| SMILES | C1=CC=NC=C1.Br |
Chemical Identifiers
| 18820-82-1 | |
| 160.014 | |
| BBFCIBZLAVOLCF-UHFFFAOYSA-N | |
| 87810 | |
| C1=CC=NC=C1.Br |
| C5H6BrN | |
| MFCD00012801 | |
| pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at | |
| pyridine;hydrobromide |
Specifications
| 18820-82-1 | |
| C5H6BrN | |
| 25 g | |
| BBFCIBZLAVOLCF-UHFFFAOYSA-N | |
| pyridine;hydrobromide | |
| 87810 | |
| ≥98.0% (HPLC,T) | |
| Pyridine Hydrobromide |
| 217°C | |
| MFCD00012801 | |
| pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at | |
| C1=CC=NC=C1.Br | |
| 160.014 | |
| 160.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes