missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide, 98%

Catalog No. AAA1931430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1931430 250 g
AAA1931418 50 g
AAA193140B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1931430 Supplier Thermo Scientific Chemicals Supplier No. A1931430

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18820-82-1 | C5H6BrN | 160.014 g/mol

Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 18820-82-1
Molecular Formula C5H6BrN
Molecular Weight (g/mol) 160.014
MDL Number MFCD00012801
InChI Key BBFCIBZLAVOLCF-UHFFFAOYSA-N
Synonym pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
PubChem CID 87810
IUPAC Name pyridine;hydrobromide
SMILES C1=CC=NC=C1.Br

Specifications

Melting Point 216°C to 220°C
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.01
Percent Purity 98%
Chemical Name or Material Pyridine hydrobromide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 242-600-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only