Learn More
Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles
Description
Pyridine is used in spectrophotometry and environmental testing.As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further, it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 42°C |
| Color | Colorless |
| Density | 0.978 g/cm3 at 20°C |
| Quantity | 100 mL |
| Flash Point | 20°C (68°F) |
| Ignition Point | 482°C (900°F) |
| Odor | Unpleasant |
| Physical Form | Liquid |
| Refractive Index | 1.51 |
| Assay Percent Range | >99.5% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H312+H332-H315-H319 |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | DOT Class: 3, Packaging Group: II |
| EINECSNumber | 203-809-9 |
| RTECSNumber | UR8400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only