missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ACS Reagent, Reagents

Catalog No. p-200020966
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Safety Coated Glass
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337781 100 mL Glass Bottle
01337780 1 L Safety Coated Glass
01337779 2.5 L Glass Bottle
01337778 4 L Glass Bottle
01337777 4 L Safety Coated Glass
01337776 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337781 Supplier Labchem Inc Supplier No. C1294000100C
Only null left

Pyridine, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym Azabenzene, Azine
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point <-29°C
Color Colorless
Boiling Point 115°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.982
Grade ACS
Physical Form Liquid
Chemical Name or Material Pyridine, ACS Reagent