missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, ACS, 99.0+%

Catalog No. p-4408286
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA19378AP 500 mL
AA19378K2 1 L
AA19378M1 4 x 1 L
AA19378K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA19378AP Supplier Thermo Scientific Chemicals Supplier No. 019378AP
Only null left

Maximum level of impurities: Evaporation residue 0.002%, H2O 0.1%, Cl 0.001%, SO4 0.001%, NH3 0.002%, Cu 5ppm, Reducing substances P.T., Solubility in water P.T. | CAS: 110-86-1 | C5H5N | 79.102 g/mol

As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane (C5H5NBH3) and pyridine-sulfur trioxide (C5H5NSO3) are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point °C to 42°C
Color Colorless
Appearance Colorless liquid
Density 0.978
Boiling Point 115°C
Quantity 500 mL
Flash Point 20°C (68°F)
Ignition Point 482°C (900°F)
Odor Unpleasant
Physical Form Liquid
Refractive Index 1.51
Assay Percent Range 99.0% min.
Beilstein 103233
Solubility Information Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight 79.1
Grade ACS
Chemical Name or Material Pyridine, For ACS analysis
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3, Packing Group: II
EINECSNumber 203-809-9
RTECSNumber UR8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only