missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, 99+%, extra pure, nonaqueous titration grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -42.0°C |
| Density | 0.9780g/mL |
| Quantity | 250 g |
| Flash Point | 17°C |
| Infrared Spectrum | Authentic |
| Packaging | EcoSafPak™ Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5085 to 1.5105 |
| Assay Percent Range | 99+% |
| Residue after Evaporation | 0.002% max. |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | Danger |
| missing translation for 'healthHazard2' | GHS P Statement Harmful in contact with skin. Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Highly flammable liquid and vapor. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only