Learn More
Pyridine, 99+%, for analysis
CAS: 110-86-1 | C5H5N | 79.1 g/mol
Supplier: Thermo Scientific Chemicals 290860010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Pyridine | |
| -42.0°C | |
| 115.0°C to 116.0°C | |
| 99% min. (GC) | |
| C5H5N | |
| MFCD00011732 | |
| 20,181 | |
| 0.978 | |
| azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| pyridine | |
| 1049 | |
| CHEBI:16227 | |
| 99+% | |
| Liquid |
| 110-86-1 | |
| 0.9780g/mL | |
| 17°C | |
| Glass bottle | |
| 1.5085 to 1.5105 | |
| 1 L | |
| 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 | |
| 15,8081 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents | |
| C1=CC=NC=C1 | |
| 79.1 | |
| 0.95 mPa.s (20°C) | |
| 79.1 | |
| Analysis |
Chemical Identifiers
| 110-86-1 | |
| 79.1 | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| 1049 | |
| pyridine |
| C5H5N | |
| MFCD00011732 | |
| azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
| CHEBI:16227 | |
| C1=CC=NC=C1 |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Danger
EINECSNumber : 203-809-9
RUO – Research Use Only