Learn More
Pyridine, 99+%, ACS reagent
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Supplier: Thermo Scientific Chemicals 418540010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Pyridine | |
| 110-86-1 | |
| 0.9780g/mL | |
| 1 L | |
| Glass Bottle | |
| 1.5085 to 1.5105 | |
| 99+% | |
| C5H5N | |
| Vapor Pressure: 20mbar at 20°C | |
| 1282 | |
| 20, 181 | |
| 15, 8081 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents | |
| C1=CC=NC=C1 | |
| 79.102 | |
| CHEBI:16227 | |
| ≥99% |
| -42.0°C | |
| Colorless | |
| 115.0°C to 116.0°C | |
| 17°C | |
| Liquid | |
| 0.002% max. | |
| 0.978 | |
| MFCD00011732 | |
| 0.95 mPa.s (20°C) | |
| 0.1% max. (K.F.) | |
| 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153 | |
| azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| pyridine | |
| 1049 | |
| 79.1 | |
| ACS Reagent |
Chemical Identifiers
| 110-86-1 | |
| 79.102 | |
| JUJWROOIHBZHMG-UHFFFAOYSA-N | |
| 1049 | |
| pyridine |
| C5H5N | |
| MFCD00011732 | |
| azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
| CHEBI:16227 | |
| C1=CC=NC=C1 |
Safety and Handling
GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 203-809-9
RTECSNumber : UR8400000
TSCA : TSCA
RUO – Research Use Only