missing translation for 'onlineSavingsMsg'
Learn More

Pyridine, 99.0% min., ACS, MilliporeSigma™

Catalog No. MPX202011
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPX202011 100 mL
MPX20206 500 mL
MPX20205 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MPX202011 Supplier Technidata Supplier No. PX202011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Color Colorless
Density 0.982g/cm3 (20°C)
Quantity 100 mL
Explosion Limit 1.7 to 10.6%(V)
Ignition Point 482°C
Physical Form Liquid
Assay Percent Range 99.0% min.
Vapor Pressure 20 hPa (20°C)
Formula Weight 79.1
Chemical Name or Material Pyridine, SeccoSolv™, Dried
Recommended Storage Store without restrictions