missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-carboxaldehyde, 97%

Catalog No. AAA1065636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065636 500 g
AAA1065614 25 g
AAA1065622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1065636 Supplier Thermo Scientific Chemicals Supplier No. A1065636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 872-85-5 | C6H5NO | 107.112 g/mol

Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-4-carboxaldehyde is an heterocyclic building block used for the synthesis of various pharmaceutical compounds, such as new 1,4-dihydropyridin-4-yl-phenoxyacetohydrazones, having anticonvulsant and anti-inflammatory properties.

Solubility
Soluble in water (20 mg/ml at 20°C), ethanol and acetone.

Notes
It is air and light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 872-85-5
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006425
InChI Key BGUWFUQJCDRPTL-UHFFFAOYSA-N
Synonym 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
PubChem CID 13389
IUPAC Name pyridine-4-carbaldehyde
SMILES C1=CN=CC=C1C=O

Specifications

Melting Point -4°C
Density 1.132
Boiling Point 198°C
Flash Point 75°C (167°F)
Refractive Index 1.544
Quantity 500 g
Beilstein 105342
Sensitivity Air and light sensitive
Solubility Information Soluble in water (20mg/ml at 20°C),ethanol and acetone.
Formula Weight 107.11
Percent Purity 97%
Chemical Name or Material Pyridine-4-carboxaldehyde
Show More Show Less
Hazard Category H227-H314-H317-H332
Hazard Statement GHS H Statement
H315-H319-H317-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 212-832-3
RTECSNumber NR9400000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only