missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine-4-boronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC04212EA
| Quantity | 10g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 1692-15-5 | |
| 122.918 | |
| pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid | |
| pyridin-4-ylboronic acid |
| C5H6BNO2 | |
| QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
| 2734379 | |
| B(C1=CC=NC=C1)(O)O |
Specifications
| Pyridin-4-ylboronic acid | |
| 97% | |
| C5H6BNO2 | |
| pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid | |
| B(C1=CC=NC=C1)(O)O | |
| 122.918 | |
| 122.92 |
| 1692-15-5 | |
| Amber Glass Bottle | |
| 10g | |
| QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
| pyridin-4-ylboronic acid | |
| 2734379 | |
| 97% |