missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-sulfonic Acid 98.0+%, TCI America™

Catalog No. P065825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P065825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P065825G Supplier TCI America Supplier No. P065825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 636-73-7
Molecular Formula C5H4NO3S
Molecular Weight (g/mol) 158.15
MDL Number MFCD00006381
InChI Key DVECLMOWYVDJRM-UHFFFAOYSA-M
Synonym 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
PubChem CID 69468
IUPAC Name pyridine-3-sulfonate
SMILES [O-]S(=O)(=O)C1=CC=CN=C1

Specifications

Melting Point 360°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 159.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-3-sulfonic Acid
RTECSNumber UT7920000
TSCA No