missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-sulfonic acid, 98+%

CAS: 636-73-7 | C5H4NO3S | 158.15 g/mol

Supplier:  Thermo Scientific Chemicals A1310114

 View more versions of this product

Catalog No. AAA1310114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pyridine-3-sulfonic acid
∼350°C (decomposition)
MFCD00006381
UN2585
Light sensitive
DVECLMOWYVDJRM-UHFFFAOYSA-M
pyridine-3-sulfonic acid
69468
≥98%
636-73-7
C5H4NO3S
25 g
120822
3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
[O-]S(=O)(=O)C1=CC=CN=C1
158.15
159.16

Chemical Identifiers

636-73-7
158.15
DVECLMOWYVDJRM-UHFFFAOYSA-M
69468
C5H4NO3S
MFCD00006381
3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
[O-]S(=O)(=O)C1=CC=CN=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID

EINECSNumber : 211-265-9

RTECSNumber : UT7920000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only