missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-sulfonic acid, 98+%

Catalog No. AAA1310136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1310136 500 g
AAA1310114 25 g
AAA1310122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1310136 Supplier Thermo Scientific Chemicals Supplier No. A1310136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 636-73-7 | C5H4NO3S | 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-73-7
Molecular Formula C5H4NO3S
Molecular Weight (g/mol) 158.15
MDL Number MFCD00006381
InChI Key DVECLMOWYVDJRM-UHFFFAOYSA-M
Synonym 3-pyridinesulfonic acid, pyridine-3-sulphonic acid, 3-pyridylsulfonic acid, 3-pyridine sulfonic acid, 3-sulfopyridine, beta-sulfopyridine, 3-pyridinesulfonicacid, pubchem9608, pyridin-3-sulfonic acid, acmc-209nh7
PubChem CID 69468
SMILES [O-]S(=O)(=O)C1=CC=CN=C1

Specifications

Melting Point ∼350°C (decomposition)
Quantity 500 g
UN Number UN2585
Beilstein 120822
Sensitivity Light sensitive
IUPAC Name pyridine-3-sulfonate
Formula Weight 159.16
Percent Purity ≥98%
Chemical Name or Material Pyridine-3-sulfonic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 211-265-9
RTECSNumber UT7920000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only