missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-carboximidamide Monohydrochloride 98.0+%, TCI America™

Catalog No. p-7135437
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21671G 1 g
P21675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21671G Supplier TCI America Supplier No. P21671G

Chemical Identifiers

CAS 7356-60-7
Molecular Formula C6H8ClN3
Molecular Weight (g/mol) 157.601
MDL Number MFCD00054326
InChI Key MKJPBOVLAZADQJ-UHFFFAOYSA-N
Synonym nicotinimidamide hydrochloride, 3-amidinopyridine hydrochloride, pyridine-3-carboximidamide hydrochloride, nicotinamidine hydrochloride, 3-pyridinecarboximidamide, monohydrochloride, nicotinamidine, hydrochloride, pyridine-3-carbonamidine hydrochloride, pyridine-3-carboxamidine hydrochloride, 3-pyridinecarboxamidine hydrochloride, pyridine-3-carboximidamide hcl
PubChem CID 202042
IUPAC Name pyridine-3-carboximidamide;hydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.Cl

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 157.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-3-carboximidamide Monohydrochloride
RTECSNumber QS4733000
TSCA No