Learn More
Pyridine-3,4-dicarboximide
CAS: 4664-01-1 | C7H4N2O2 | 148.12 g/mol
$85.26 - $806.74
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3175806
|
Thermo Scientific Chemicals
H3175806 |
5 g |
Each for $85.26
|
|
|||||
|
AAH3175814
|
Thermo Scientific Chemicals
H3175814 |
25 g |
Each for $285.99
|
|
|||||
|
AAH3175822
|
Thermo Scientific Chemicals
H3175822 |
100 g |
Each for $806.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4664-01-1 | |
| MFCD00013439 |
Specifications
| 4664-01-1 | |
| C7H4N2O2 | |
| 5 g | |
| SJSABZBUTDSWMJ-UHFFFAOYSA-N | |
| pyrrolo[3,4-c]pyridine-1,3-dione | |
| 72926 | |
| Pyridine-3,4-dicarboximide |
| 230°C to 232°C | |
| MFCD00013439 | |
| 3,4-pyridinedicarboximide, cinchomeronimide, 3,4-pyridinecarboximide, 1h-pyrrolo 3,4-c pyridine-1,3 2h-dione, 2h-pyrrolo 3,4-c pyridine-1,3-dione, pyrrolo 3,4-c pyridine-1,3-dione, cinchomeronic acid imide, 4-azaphthalimid, pyridine-3,4-dicarboximide, 1h,2h,3h-pyrrolo 3,4-c pyridine-1,3-dione | |
| O=C1NC(=O)C2=CN=CC=C12 | |
| 148.12 | |
| 148.12 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : US7966000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only