missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3,4-dicarboximide

Catalog No. AAH3175822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3175822 100 g
AAH3175806 5 g
AAH3175814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3175822 Supplier Thermo Scientific Chemicals Supplier No. H3175822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4664-01-1 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4664-01-1
MDL Number MFCD00013439

Specifications

Melting Point 230°C to 232°C
Molecular Formula C10H11O4
Quantity 100 g
Synonym 3,5-dimethoxyphenylacetic acid, 2-3,5-dimethoxyphenyl acetic acid, 3,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,5-dimethoxy, 3,5-dimethoxyphenylaceticacid, 3,5-dimethoxybenzeneacetic acid, 3,5-dimethoxy-phenylacetic acid, 3,5-dimethoxy phenylacetic acid, 3,5-dimethoxy phenyl acetic acid
InChI Key FFPAFDDLAGTGPQ-UHFFFAOYSA-M
SMILES COC1=CC(=CC(=C1)CC(=O)O)OC
IUPAC Name 2-(3,5-dimethoxyphenyl)acetic acid
Molecular Weight (g/mol) 195.20
PubChem CID 138316
Formula Weight 148.12
Chemical Name or Material Pyridine-3,4-dicarboximide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber US7966000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only